missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bohlender™ BOLA™ Eco Stopcock Plugs without Bore, Taper 1:10, PP
Click to view available options
Material:
Polypropylene
Unit Size:
Pack of 20
Description
- Without bore
- Smooth surface
- Grip and nut made of polypropylene
- Washer included
Specifications
Specifications
| Description | Taper 1:10 |
| Material | Polypropylene |
| Closure Size | NS 12.5 x 27 mm |
| For Use With (Equipment) | Thread M 8 |
| Color | Red |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction