Learn More
Bohlender™ BOLA™ Center Magnetic Stirring Bars
Product Code. 17107054
Click to view available options
Length (Metric):
20 mm
30 mm
40 mm
50 mm
Diameter (Metric):
7 mm
8 mm
Length (English):
0.79 in.
1.2 in.
1.6 in.
2 in.
Diameter (English):
0.28 in.
0.31 in.
Unit Size:
Pack of 10
Pack of 3
Product Code. 17107054
Brand: Bohlender™ C36720
PTFE-coated center magnetic stirring bars with conical endings.
  • PTFE-encapsulated magnetic core (Alnico 5)
  • Conically tapered ends
  • Universal chemical resistance
Specifications
Diameter (English) 0.28 in.
Length (English) 0.79 in.
Coating PTFE
Diameter (Metric) 7 mm
Length (Metric) 20 mm
Shape Conically Tapered Ends