missing translation for 'onlineSavingsMsg'
Learn More

Bodipy Cyclopamine (>85%), TRC

Product Code. 30340956
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30340956 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340956 Supplier Toronto Research Chemicals Supplier No. TRCB6748001MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bodipy Cyclopamine
CAS 334658-24-1
Molecular Formula C49H70BF2N5O4
Quantity 1 mg
Synonym Boron, [5-[(3,5-dimethyl-2H-pyrrol-2-ylidene-κN)methyl]-N-[6-[[2-[(3S,3'R,3'aS,6'S,6aS,6bS,7'aR,2'R,11aS,11bR)-1,2,3,3',3'a,4,5',6,6',6a,6b,7,7',7'a,8,11,11a,11b-octadecahydro-3-hydroxy-3',6',10,11b-tetramethylspiro[9H-benzo[a]fluorene-9,2'(4'H)-furo[3,2-b]pyridin]-4'-yl]ethyl]amino]-6-oxohexyl]-1H-pyrrole-2-propanamidato-κN1]difluoro-, (T-4)-, (T-4)-[5-[(3,5-Dimethyl-2H-pyrrol-2-ylidene-κN)methyl]-N-[6-[[2-[(3S,3'R,3'aS,6'S,6aS,6bS,7'aR,2'R,11aS,11bR)-1,2,3,3',3'a,4,5',6,6',6a,6b,7,7',7'a,8,11,11a,11b-octadecahydro-3-hydroxy-3',6',10,11b-tetramethylspiro[9H-benzo[a]fluorene-9,2'(4'H)-furo[3,2-b]pyridin]-4'-yl]ethyl]amino]-6-oxohexyl]-1H-pyrrole-2-propanamidato-κN1]difluoroboron, BODIPY-cyclopamine, Bodipy cyclopamine
InChI Formula InChI=1S/C49H70BF2N5O4/c1-30-24-44-47(34(5)49(61-44)20-18-39-40-15-11-35-26-38(58)17-19-48(35,6)42(40)28-41(39)33(49)4)55(29-30)23-22-54-45(59)10-8-7-9-21-53-46(60)16-14-36-12-13-37-27-43-31(2)25-32(3)56(43)50(51,52)57(36)37/h11-13,25,27,30,34,38-40,42,44,47,58H,7-10,14-24,26,28-29H2,1-6H3,(H,53,60)(H,54,59)/t30?,34?,38-,39?,40?,42?,44+,47?,48?,49?/m0/s1
SMILES C[C@H]1C[C@H]2O[C@]3(CC[C@H]4[C@@H]5CC=C6C[C@@H](O)CC[C@]6(C)[C@H]5CC4=C3C)[C@H](C)[C@@H]2N(CCNC(=O)CCCCCNC(=O)CCc7ccc8C=C9C(=CC(=[N+]9[B-](F)(F)n78)C)C)C1
IUPAC Name N-[2-[(3S,7'aR)-3-hydroxy-3',6',10,11b-tetramethylspiro[2,3,4,6,6a,6b,7,8,11,11a-decahydro-1H-benzo[a]fluorene-9,2'-3,3a,5,6,7,7a-hexahydrofuro[3,2-b]pyridine]-4'-yl]ethyl]-6-[3-(2,2-difluoro-10,12-dimethyl-1-aza-3-azonia-2-boranuidatricyclo[7.3.0.03,7]dodeca-3,5,7,9,11-pentaen-4-yl)propanoylamino]hexanamide
Molecular Weight (g/mol) 841.92
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.