Learn More
Macherey-Nagel™ Nanaocolor™ BOD5-TT Accessory Kit
Required when using the Nanocolor BOD5 Tube Test for photometric determination. Macherey-Nagel™ Nanaocolor™ BOD5-TT Accessory Kit includes: electric air pump, lid, 2 aerating bricks, 1000mL storage vessel, 1m flexible tube, and 2 x 40mL reaction vessels.
Brand:  Macherey-Nagel™ 916925
Product Code. 10729372
  • 98.35€ / Pack of 1
Only null left
In Stock
Description
The Nanaocolor™ BOD5-TT enables simple determination of the biochemical oxygen demand after 5 days (BOD5) of undiluted samples without using a control in accordance with DIN EN 1899-2H52.
  • The incubation of the samples is directly carried out in test tubes
  • Determination of DO based on the Winkler procedure, DIN EN 25813-G21
  • Reaction basis according to DIN EN 1899-1-H51 and DIN EN 25813-G21
Specifications
Specifications
O2
Tube Test
NANOCOLOR™ BOD5-TT (man. no. 985 825)
1 Pack
Winkler
DIN EN 1899-2H52
Photometric determination of biochemical oxygen demand after 5 days
electric air pump, lid with bore-hole, 2 aerating bricks, 1000mL storage vessel, 1m flexible tube, 2 x 40mL reaction vessels
0.5 - 300mg/L
BOD5-TT Accessories Set