Learn More
Boc-His-OMe, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 2488-14-4 |
| Molecular Formula | C12H19N3O4 |
| MDL Number | MFCD00167635 |
| Quantity | 5 g |
| SMILES | O=C(OC)[C@@H](NC(OC(C)(C)C)=O)CC1=CN=CN1 |
| Molecular Weight (g/mol) | 269.30 |
| Percent Purity | 97% |
| Chemical Name or Material | Boc-His-OMe |