Learn More
Boc-D-serine, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Boc-D-serine |
| CAS | 6368-20-3 |
| Molecular Formula | C8H15NO5 |
| Quantity | 10 g |
| Synonym | N-[(1,1-dimethylethoxy)carbonyl]-D-serine, D-N-Carboxy-serine N-tert-Butyl Ester, (R)-2-(tert-Butoxycarbonylamino)-3-hydroxypropanoic Acid, (R)-2-(tert-Butoxycarbonylamino)-3-hydroxypropanoic Acid, N-Boc-D-serine, N-tert-Butoxycarbonyl-D-serine, tert-(Butoxycarbonyl)-D-serine |
| InChI Formula | InChI=1S/C8H15NO5/c1-8(2,3)14-7(13)9-5(4-10)6(11)12/h5,10H,4H2,1-3H3,(H,9,13)(H,11,12)/t5-/m1/s1 |
| SMILES | CC(C)(C)OC(=O)N[C@H](CO)C(=O)O |
| IUPAC Name | (2R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
| Molecular Weight (g/mol) | 205.21 |
| Formula Weight | 205.095 |