Learn More
Helios Italquartz Boats with Ring Handle, Translucent
Boats with ring handle constructed of translucent, durable, resistant, and chemically inert quartz glass.
Brand:  Helios Italquartz 62F00105
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15888024
  • 124.00€ / Each
Estimated Shipment: 19-06-2024
Description
  • Translucent
  • Include ring handle
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Specifications
Flat
Quartz
Boat with Ring Handle
76/19 mm
11 mm
Translucent
Boat
76 mm x 11 mm
11 mm