Learn More
Helios Italquartz Boats with Ring Handle, Translucent
Boats with ring handle constructed of translucent, durable, resistant, and chemically inert quartz glass.
123.00€ - 255.00€
Spécification
Color | Translucent |
---|---|
Material | Quartz |
Shape | Boat |
Type | Boat with Ring Handle |
Code produit | Marque | Dimensions (L x W) | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Dimensions (L x W) | Prix | Quantité et disponibilité | |||||
15878024
|
Helios Italquartz
62F00104 |
76 mm x 10 mm |
123.00€
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15888024
|
Helios Italquartz
62F00105 |
76 mm x 11 mm |
124.00€
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15898024
|
Helios Italquartz
62F00107 |
120 mm x 15 mm |
181.00€
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15808034
|
Helios Italquartz
62F00108 |
180 mm x 15 mm |
233.00€
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15818034
|
Helios Italquartz
62F00109 |
143 mm x 17 mm |
255.00€
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
Description
- Translucent
- Include ring handle
- Chemically inert
- Excellent electrical insulation
- Excellent strength and durability
- Exceptional thermal stability compared to glass
Spécification
Translucent | |
Quartz | |
Boat | |
Boat with Ring Handle |