Learn More
Helios Italquartz Boats with Ring Handle, Translucent
Boats with ring handle constructed of translucent, durable, resistant, and chemically inert quartz glass.
Brand: Helios Italquartz 62F00104
Additional Details : Weight : 0.20000kg
Description
- Translucent
- Include ring handle
- Chemically inert
- Excellent electrical insulation
- Excellent strength and durability
- Exceptional thermal stability compared to glass
Specifications
Flat | |
Quartz | |
Boat with Ring Handle | |
76/16 mm | |
10 mm |
Translucent | |
Boat | |
76 mm x 10 mm | |
10 mm |