missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMX-IN-1

Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
BMX-IN-1 is a potent and irreversible BMX inhibitor (IC50 = 25 nM). Covalently modifies Cysteine 496. Demonstrates selectivity over other Tec family members with identically positioned reactive cysteine residue in vitro. Reduces cell number and induces apoptosis in prostate cancer cell lines. Also inhibits proliferation and reduces viability in HeLa and SiHa cell lines.
Specifications
Specifications
| Chemical Name or Material | N-[2-Methyl-5-[9-[4-[(methylsulfonyl)amino]phenyl]-2-oxobenzo[h]-1,6-naphthyridin-1(2H)-yl]phenyl]-2-propenamide |
| CAS | 1431525-23-3 |
| Assay Percent Range | >98% |
| Quantity | 5 mg |
| Molecular Formula | C29H24N4O4S |
| Formula Weight | Observed MW: 524.59 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction