Learn More
IKA™ BMT-50-G Tubes
Product Code. 11876810 Shop All IKA Products
Change view
Click to view available options
For Use With (Equipment):
BMT-50-G
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11876810 BMT-50-G Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11876810 Supplier IKA™ Supplier No. 0003699800
Only null left
Specifications
Product Type BMT-50-G Tube
Volume Range (Metric) 15 to 50 mL
For Use With (Application) Dry milling of dry and brittle samples, cell maceration, processing of materials mixed with fluids
For Use With (Equipment) BMT-50-G
Packaging Quantity 10/Pk.