missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BMS 309403
Description
BMS 309403 is a potent and selective fatty acid binding protein 4, adipocyte (FABP4) inhibitor (Ki values are <2, 250 and 350 nM for FABP4, FABP3 and FABP5 respectively). Decreases fatty acid uptake in adipocytes in vitro and reduces atherosclerotic lesion area in a mouse model of atherosclerosis. Reduces blood glucose levels and increases insulin sensitivity in a mouse model of obesity. Orally active.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | BMS 309403 |
| Purity | >98% |
| Molecular Weight (g/mol) | 474.55 |
| Inhibitors | BMS 309403 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?