Learn More
Brady™ BMP71 BMP61 M611 Continuous Metallized Polyester Asset Tracking Labels
Click to view available options
Color:
Gray
Unit Size:
1 roll
Description
- Material is UL, CSA & AGA approved with its respective ribbon
- See the Recommended Ribbon Series for the appropriate ribbon to use
- Can also be printed on the BMP™71 Label Printer provided the roll adapter (M71-ADAPT) is used
Specifications
Specifications
| Adhesive Type | Permanent Acrylic |
| Color | Gray |
| Finish | Matte |
| Material | Polyester (B-428) |
| Print Technology | Thermal Transfer |
| For Use With (Equipment) | BMP61, BMP71 (with Media Adapter), M611 |
| Dimensions (LxW) | 15.24 m x 12.7 mm |