Learn More
Brady™ BMP71 BMP61 M611 Clear Polyester Asset Tracking Labels
Product Code. 11539033
Click to view available options
Dimensions (LxW):
15.24 m x 25.4 mm
15.24 m x 48.26 mm
15.24 m x 9.53 mm
Unit Size:
1 roll
Product Code. 11539033
Brand: Brady™ 018619
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Labels for the BMP61 and M611 Thermal Transfer Portable Printers.
  • Material is UL & CSA approved with its respective ribbon
  • See the Recommended Ribbon Series for the appropriate ribbon to use
  • Can also be printed on the BMP™71 Label Printer provided the roll adapter (M71-ADAPT) is used
Specifications
Adhesive Type Permanent Acrylic
Color Transparent
Finish Gloss
Material Polyester (B-430)
Print Technology Thermal Transfer
For Use With (Equipment) BMP61, BMP71 (with Media Adapter), M611
Dimensions (LxW) 15.24 m x 25.4 mm