Learn More
Brady™ BMP61 M611 Self-Laminating Polyester Labels
GREENER_CHOICE
Product Code. 17130411
Click to view available options
Product Type:
Printer Label
Printer Tapes
Size (English):
0.375 in. x 50 ft. (W x L)
0.5 in. x 50 ft. (W x L)
0.5 x 1.65 in. (W x H)
1 in. x 50 ft. (W x L)
1 x 1.75 in. (W x H)
1.125 x 0.375 in. (W x H)
1.125 x 0.5 in. (W x H)
1.5 x 2.5 in. (W x H)
Unit Size:
Each
Pack of 100
Pack of 500
Product Code. 17130411
Brand: Brady™ 150862
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
A clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
  • Good smudge resistance, solvent resistance and high/low-temperature performance
  • Service temperature of -320 to 230° F (-196 to 110° C)
  • Provided in a self-laminated format with a white printable zone and translucent overlaminating area to protect text
Specifications
Adhesive Type Acrylic
Color White
Material Polyester
Print Technology Thermal Transfer
Product Type Printer Tapes
Size (English) 0.375 in. x 50 ft. (W x L)
Dimensions (LxW) 50 ft. x 0.375 in.
Length (English) 50 ft.
Width (English) 0.375 in.