Learn More
Brady™ BMP61 Label Printer with AZERTY Keyboard and Workstation PWID Suite
Product Code. 16389141
Click to view available options
Unit Size:
Each
Product Code. 16389141
Brand: Brady™ 198633
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The rugged, thermal transfer BMP61 Label Printer can print sleeves, self-laminating labels, raised profile labels, tags and other identification solutions in a vast range of sizes.
Rugged and reliable, the drop-tested BMP™61 Label Printer is equipped with molded rubber bumpers for extra protection.
  • Fast printing speed
  • Handles die-cut, continuous and custom labels up to 50.8 mm (also in bulk format)
  • Large color touchscreen with indicators allows for easy checking to see which label is installed and how many labels you have left
  • Read spreadsheets directly from external USB-memory and integrate the data into labels ready for printing
  • Wi-Fi version available
  • Compatible with mobile apps- create or load label templates with your smartphone and send them to your printer
  • AZERTY keyboard
  • Comes with Brady™ Workstation PWID Suite
Specifications
Display Type Color LCD
Print Technology Thermal Transfer
Width (English) 4 in.
Width (Metric) 107 mm
Height (English) 13 in.
Height (Metric) 328 mm
Depth (English) 4 in.
Depth (Metric) 106.4 mm
Autoclavable Not Autoclavable
Sterility Non-sterile
Color Capability Monochrome
Includes Printer, USB Cable, Quick Start Guide, Battery Pack, AC Adapter, CD with Driver, Product CD, Brady™ Workstation Product & Wire Identification Suite, Roll of PTL-19-423, and M61-R6010 Ribbon
Languages Bulgarian, Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Japanese, Norwegian, Polish, Portuguese, Romanian, Russian, Slovakian, Slovenian, Spanish, Swedish, Turkish
Memory 512 MB RAM, 256 MB Flash
Model BMP61
Print Speed 33.8 mm/s
Resolution 300 dpi
Connector Types USB
Temperature Range (Metric) 5°C to 49°C
Weight (English) 3 lb.
Weight (Metric) 1.4 kg
For Use With (Application) Barcode and Inventory Labeling, Hazard Identification, Laboratory Labeling
Warranty 2 Year
Voltage 110/240 V