Learn More
Tocris Bioscience™ BML 111, Tocris Bioscience™
GREENER_CHOICE
FPR2 (lipoxin A4 receptor) agonist
Brand:  Tocris Bioscience™ 5488/10
Encompass
Product Code. 15776876
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
BML 111 is a FPR2 (lipoxin A4 receptor) agonist. Suppresses tumor-related angiogenesis and reduces tumor growth in hepatoma H22 cell bearing mice. Also protects against hemorrhagic shock-induced acute lung injury in rats. Lipoxin A4 analog.