Learn More
Tocris Bioscience™ BML 111, Tocris Bioscience™
FPR2 (lipoxin A4 receptor) agonist
Brand: Tocris Bioscience™ 5488/10
This item is not returnable.
View return policy
Description
BML 111 is a FPR2 (lipoxin A4 receptor) agonist. Suppresses tumor-related angiogenesis and reduces tumor growth in hepatoma H22 cell bearing mice. Also protects against hemorrhagic shock-induced acute lung injury in rats. Lipoxin A4 analog.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only.