missing translation for 'onlineSavingsMsg'
Learn More

Melford Bluo-Gal

Product Code. p-200086999
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
This item is not returnable. View return policy

Product Code. 30018108

missing translation for 'mfr': Melford B725001.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chromogenic substrate for B-D-Galactosidase.

Chromogenic substrate for B-D-Galactosidase. Produces a deeper blue water insoluble color than X-GAL.

Specifications

Quantity 1 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.