Learn More
3M™ Blue SMS Based Fabric Category I Protective Coveralls
Click to view available options
:
Size: X-Large
Specifications
Specifications
| Product Type | General Purpose Coveralls |
| Gender | Unisex |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Hood Type | 2-panel |
| Seam Type | Serged |
| Sterility | Non-sterile |
| Show More |