Learn More
mb2 Chair on Castors - Blue Fabric Upholstered - Asynchronous Mechanism
Product Code. 11977264
Click to view available options
:
Height: 700mm
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Product Code. 11977264
Brand: mb2 AT3XB2D1NE2D6016
Only null left
Add to basket
Edge
Add to basket
You may only order up to the available stock. For more details call customer services on +351 21 425 33 50.
Chair on castors - Mid lift - Asynchronous mecanism - blue fabric upholstered
  • Chair in fabric blue
  • Seat and back in blue fabric upholstered
  • Asynchronous mecanism, adjustable seat and backrest
  • Pneumatic cylinder Mid
  • 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
  • Hard floor castors 50 mm
Specifications
Product Type Contact Seat Chair
Color Blue
Adjustable (Height) Yes
Adjustable Height
Back Style Ergonomic
Height (Metric) 700 mm
Holds 5 Wheels
Seating Height 472 to 710 mm
Weight (Metric) 10.40 kg
With Armrest No
With Backrest Yes
With Casters Yes
Footrest No
With Lumbar Support No
With Tilt Yes
Material Fabric
Seating Type Medium Bench Height