Learn More
mb2 Chair on Castors - Blue Fabric Upholstered - Asynchronous Mechanism
Product Code. 11957264
Click to view available options
:
Height: 540mm
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11957264
missing translation for 'mfr': mb2 AT3XB1D1NE2D6016
Bara null kvar
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy
Chair on castors - Normal lift - Asynchronous mecanism - blue fabric upholstered
  • Chair in fabric blue
  • Seat and back in blue fabric upholstered
  • Asynchronous mecanism, adjustable seat and backrest
  • Pneumatic cylinder low
  • 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
  • Hard floor castors 50 mm
Specifications
Product Type Contact Seat Chair
Color Blue
Adjustable (Height) Yes
Adjustable Height
Back Style Ergonomic
Height (Metric) 540 mm
Holds 5 Wheels
Seating Height 440 to 560 mm
Weight (Metric) 10.20 kg
With Armrest No
With Backrest Yes
With Casters Yes
Footrest No
With Lumbar Support No
With Tilt Yes
Material Fabric
Seating Type Desk Height