Learn More
Vitlab™ Blue Raised Scale Graduated Beaker
Product Code. 10602821
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
3000 mL
5000 mL
Unit Size:
Each
Pack of 12
Pack of 24
Pack of 6
Product Code. 10602821
Brand: Vitlab™ 445081
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use these polypropylene beakers for a variety of general laboratory applications. VITLAB™ Blue Raised Scale Graduated Beakers feature a stable handle easy-grip handle and an easily readable, embossed blue scale.
  • Highly transparent polypropylene beaker
  • To preserve markings, do not clean at temperatures exceeding 60°C
Specifications
Color Transparent
Diameter (Metric) Outer 210 mm
Capacity (Metric) 5000 mL
Material Polypropylene
Height (Metric) 270 mm
Quantity 6 Pk.
Type Beaker
Autoclavable Autoclavable
Graduated Yes
Certifications/Compliance DIN EN 285
Scale Blue Raised Scale