Learn More
3M™ Protective Coverall 4532+, Blue
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Pack of 20
Description
- Unique liquid repellency with oil and alcohol resistance material treatment
- Soft and strong multi-layered 52 gsm SSMMS for strong performance
- Made from super breathable SSMMS non-woven for effective release of body heat and moisture and reduced heat stress
- 3 panel hood, reinforced triangular gusset panel, 2-way zip and storm flap with sealable tape, elasticated back and ankles to better freedom of movement
- Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
- EN 1073-2 nuclear particulate hold out
- Knitted cuffs and elastic waist and ankles for a better fit and freedom of movement
Specifications
Specifications
| Product Type | Protective Coveralls |
| Size | 4X-Large |
| Gender | Unisex |
| Material | SSMMS |
| Color | Blue |
| Disposable | Single-use |
| Waist Style | Elastic |
| Cuff Style | Knit |
| Closure Type | Front Zipper |
| Sterility | Non-sterile |
| Show More |