Learn More
Fisherbrand™ Color-coded Octahedral PTFE Stir Bar
Product Code. 11888912 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
13 mm
15 mm
22 mm
25 mm
38 mm
51 mm
75 mm
Diameter (Metric):
8 mm
13 mm
Color:
Blue
Red
Yellow
Unit Size:
Pack of 10
Pack of 5
Product Code. 11888912
Use when quick visual identification is highly important. Fisherbrand™ Color-coded Octahedral PTFE Stir Bar is available in three colors: blue, red, and yellow.
  • Color-coded, octagonal stirrer bars
  • For use where identification is of prime importance
  • Note: Colored PTFE coatings are not as inert as pure PTFE
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color Blue
Description PTFE encapsulated Alnico V magnet core
Diameter (English) 0.5 in.
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Length (English) 3 in.
Diameter (Metric) 13 mm
Length (Metric) 75 mm
Quantity 2/Pk.
Shape Octahedral