Learn More
Melford Blotto-Tween 20
Product Code. 30018131
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018131 500 mL 500mL
30018130 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30018131 Supplier Melford Supplier No. B81000500.0
 Restricted Item
Estimated Shipment: 15-12-2026
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Solution contains:
50mM TRIS-HCl
150mM NaCl
3% BSA
0.05% Tween-20
0.02% Sodium Azide
Optimized for use in western blotting to minimize non-specific binding of antibodies.
pH: 7.4 ± 0.2
Specifications
Quantity 500 mL