missing translation for 'onlineSavingsMsg'
Learn More

Melford Blotto-Tween 20

Product Code. 30018130
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
This item is not returnable. View return policy

Product Code. 30018130

Brand: Melford B810001000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

Solution contains:

50mM TRIS-HCl

150mM NaCl

3% BSA

0.05% Tween-20

0.02% Sodium Azide

Optimized for use in western blotting to minimize non-specific binding of antibodies.

pH: 7.4 ± 0.2

Specifications

Quantity 1 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.