Learn More
Benchmark Scientific™ Block for Benchmark MultiTherm
Click to view available options
No. of Places:
6
Unit Size:
Each
Description
- Accommodates 6 x 50 mL tubes
- Protective shield protects users from burns
- Fits MultiTherm H and HC Models
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Chamber Material | Aluminum |
| Holds | 6 x 50 mL Tubes |
| Material | Aluminum |
| No. of Places | 6 |
| Type | Block |