Learn More
Benchmark Scientific™ Block for Benchmark MultiTherm
Product Code. 17904412
Click to view available options
No. of Places:
6
Unit Size:
Each
Product Code. 17904412
Brand: Benchmark Scientific™ H5000500
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This block holds 6 x 50 mL tubes. Made of aluminum, it is covered with a protective shield. Fits Benchmark MultiTherm.
  • Accommodates 6 x 50 mL tubes
  • Protective shield protects users from burns
  • Fits MultiTherm H and HC Models
Specifications
Autoclavable Not Autoclavable
Chamber Material Aluminum
Holds 6 x 50 mL Tubes
Material Aluminum
No. of Places 6
Type Block