Learn More
BRAND™ Blaubrand™ One-Mark Bulb Pipettes, with Certificate
Product Code. 15801475 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
2 mL
2.5 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15801475 2 mL Each
10029529 2.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15801475 Supplier BRAND™ Supplier No. 929703
Only null left
Add to basket
Add to basket
Unique class AS volumetric pipettes made of Soda-lime glass with 1 marking and blue graduation. Delivered with individual conformity certificate.
  • Volumetric pipettes, class AS, Soda-lime glass, 1 marking, blue graduation
  • Blaubrand™ DIN EN ISO 648
  • DE-M marking with batch number
  • Graduations and inscriptions in highly contrasting blue enamel Calibrated to deliver (EX)
  • All Blaubrand™ volumetric pipettes are supplied with batch number and one batch certificate per manufacturer's outer pack (up to 2ml: 12 cylinders, above 2ml: 6 cylinders per pack)
  • Delivered with individual conformity certificate
TRUSTED_SUSTAINABILITY
Specifications
Calibration To Deliver
Certifications/Compliance DIN EN ISO 648
Class AS
Material AR-GLAS
Min. Flow Time 5 sec.
Tolerance 0.01 mL
Color Orange
Volume (Metric) 2 mL
Length (Metric) 330 mm
Type One-mark Bulb Pipet