missing translation for 'onlineSavingsMsg'
Learn More

Melford Blasticidin S HCL Powder

Product Code. 30018072
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018072 100 mg 100mg
30018071 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018072 Supplier Melford Supplier No. B122000.1
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Blasticidin S hydrochloride powder is a commonly used antibiotic in the life science marketplace.

Blasticidin S hydrochloride powder is a commonly used antibiotic in the life science marketplace. It is primarily employed as a selective agent in cell culture systems and molecular biology applications.

Blasticidin S is derived from Streptomyces griseochromogenes and is effective against a wide range of organisms, including bacteria, yeast, fungi, and certain higher eukaryotic cells. It works by inhibiting protein synthesis in cells, specifically targeting the 80S ribosome and interfering with the peptidyl transferase activity.

In the life science marketplace, Blasticidin S hydrochloride powder is frequently used in cell culture research and genetic engineering. It is added to growth media to select for cells that have successfully incorporated a Blasticidin S resistance gene, typically as part of a recombinant DNA construct or plasmid vector.

This antibiotic selection system allows researchers to identify and maintain only those cells that have taken up the desired genetic material. The selective nature of Blasticidin S hydrochloride powder makes it an essential tool in molecular biology techniques such as gene expression studies, protein production, and stable cell line generation. It enables the establishment of cell cultures with stable gene expression, allowing researchers to study gene function and protein interactions.

Specifications

Chemical Name or Material Blasticidin S Hydrochloride Powder
CAS 3-9-3513
Quantity 100 mg
Molecular Formula C17H26N8O5-HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.