Learn More
mb2 Chair on Castors - Black Fabric Upholstered - Asynchronous Mechanism
Product Code. 11917274
Click to view available options
:
Height: 660mm
Product Code. 11917274
Brand: mb2 AT3XB2D1NE2D8033
Chair on castors - Mid lift - Asynchronous mecanism - black fabric upholstered
  • Chair in fabric black
  • Seat and back in blue fabric upholstered
  • Asynchronous mecanism, adjustable seat and backrest
  • Pneumatic cylinder Mid
  • 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
  • Hard floor castors 50 mm
Specifications
Product Type Contact Seat Chair
Color Black
Adjustable (Height) Yes
Adjustable Height
Back Style Ergonomic
Height (Metric) 660 mm
Holds 5 Wheels
Seating Height 472 to 710 mm
Weight (Metric) 10.40 kg
With Armrest No
With Backrest Yes
With Casters Yes
Footrest No
With Lumbar Support No
With Tilt Yes
Material Fabric
Seating Type Medium Bench Height