Learn More
asecos™ Black PE Drum Stand
Product Code. 17876436 Shop All Asecos Products
Change view
Click to view available options
Material:
Polyethylene
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
17876436 Polyethylene Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17876436 Supplier asecos™ Supplier No. 9875
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
The Black PE Drum Stand is suitable for 60 and 200 liter drums with 180° rotation.
  • Total load capacity: 400 kg.
  • Dimensions W x D x H (mm) exterior: 620 x 473 x 390.
Specifications
Color Black
Material Polyethylene
Type Drum Rack
Length (English) 18.62 in.
Length (Metric) 473.00 mm
Width (English) 18.62 in.
Width (Metric) 473.00 mm
Height (English) 15.35 in.
Height (Metric) 390.00 mm
Depth (English) 24.41 in.
Depth (Metric) 620.00 mm
Weight (English) 19.84 lb.
Weight (Metric) 9 kg