missing translation for 'onlineSavingsMsg'
Learn More

Biuret, 97%

Product Code. p-7043447
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488779 25 g 25g
11330244 100 g 100g
11340244 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15488779 Supplier Thermo Scientific Alfa Aesar Supplier No. L00812.14
Add to basket
This item is not returnable. View return policy

CAS: 108-19-0 | C2H5N3O2 | 103.081 g/mol

Biuret is used in protein assay. It is also used as Catalytic agent, Petrochemical additive. It is used in organic synthesis. Biuret (Carbamoylurea) was used as a source of dietary nitrogen in purified diets for steers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biuret is used in protein assay. It is also used as Catalytic agent, Petrochemical additive. It is used in organic synthesis. Biuret (Carbamoylurea) was used as a source of dietary nitrogen in purified diets for steers.

Solubility
Soluble in methanol. Partially soluble in cold water, diethyl ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-19-0
Molecular Formula C2H5N3O2
MDL Number MFCD00007946
InChI Key OHJMTUPIZMNBFR-UHFFFAOYSA-N
Synonym biuret, allophanamide, imidodicarbonic diamide, carbamylurea, ureidoformamide, dicarbonimidic diamide, allophanic acid amide, allophanimidic acid, dicarbamylamine, isobiuret
PubChem CID 7913
ChEBI CHEBI:18138
IUPAC Name carbamoylurea

Specifications

Melting Point ∼189°C (decomposition)
Density 1.46
Odor Odorless
Linear Formula H2NCONHCONH2
Quantity 25 g
Beilstein 1703510
Merck Index 14,1309
Solubility Information Soluble in methanol. Partially soluble in cold water,diethyl ether.
SMILES NC(=O)NC(N)=O
Molecular Weight (g/mol) 103.08
Formula Weight 103.08
Percent Purity 97%
Chemical Name or Material Biuret
Show More Show Less
compliance-icons
Product Identifier
  • Biuret
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Imidodicarbonic diamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.