Learn More
Biuret-15N3, TRC
Product Code. 30333803
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30333803 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30333803 Supplier Toronto Research Chemicals Supplier No. TRCB59150225MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Biuret-15N3
CAS 287484-46-2
Molecular Formula C2H515N3O2
Quantity 25 mg
Synonym Imidodicarbonic-15N di(amide-15N), Allophanamide-15N3, Allophanic Acid Amide-15N3, Allophanimidic Acid-15N3, Carbamylurea-15N3, Desmodeyn 75-15N3, Dicarbamylamine-15N3, HO-1-15N3, HO 1 (Biuret)-15N3, Isobiuret-15N3, N 75-15N3, NSC 8020-15N3, (aminocarbonyl)urea-15N3, Ureidoformamide-15N3, Imidodicarbonic diamide-15N3
InChI Formula InChI=1S/C2H5N3O2/c3-1(6)5-2(4)7/h(H5,3,4,5,6,7)/i3+1,4+1,5+1
SMILES [15NH2]C([15NH]C([15NH2])=O)=O
IUPAC Name (15N)azanylcarbonylurea
Molecular Weight (g/mol) 106.06
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.