Learn More
Biuret-15N3, TRC
Specifications
Specifications
| Chemical Name or Material | Biuret-15N3 |
| CAS | 287484-46-2 |
| Molecular Formula | C2H515N3O2 |
| Quantity | 10 mg |
| Synonym | Imidodicarbonic-15N di(amide-15N), Allophanamide-15N3, Allophanic Acid Amide-15N3, Allophanimidic Acid-15N3, Carbamylurea-15N3, Desmodeyn 75-15N3, Dicarbamylamine-15N3, HO-1-15N3, HO 1 (Biuret)-15N3, Isobiuret-15N3, N 75-15N3, NSC 8020-15N3, (aminocarbonyl)urea-15N3, Ureidoformamide-15N3, Imidodicarbonic diamide-15N3 |
| InChI Formula | InChI=1S/C2H5N3O2/c3-1(6)5-2(4)7/h(H5,3,4,5,6,7)/i3+1,4+1,5+1 |
| SMILES | [15NH2]C([15NH]C([15NH2])=O)=O |
| IUPAC Name | (15N)azanylcarbonylurea |
| Molecular Weight (g/mol) | 106.06 |