Learn More
Bisphenol F Diglycidyl Ether, TRC
Specifications
Specifications
| Chemical Name or Material | Bis(4-glycidyloxyphenyl)methane |
| CAS | 2095-03-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19 H20 O4 |
| Quantity | 50 mg |
| Synonym | BFDGE, Methane, bis[p-(2,3-epoxypropoxy)phenyl]- (7CI,8CI), 2,2'-[Methylenebis(4,1-phenyleneoxymethylene)]bis[oxirane], 4,4'-Methylenebisphenol diglycidyl ether, 4,4'-Methylenediphenol diglycidyl ether, Bis(4-(oxiran-2-ymethoxy)phenyl)methane, Bis(4-glycidyloxyphenyl)methane, Bis(4-hydroxyphenyl)methane diglycidyl ether, Bis(p-hydroxyphenyl)methane diglycidyl ether, Bisphenol F diglycidyl ether, p,p-BFDGE, para-para-BFDGE |
| InChI Formula | InChI=1S/C19H20O4/c1-5-16(20-10-18-12-22-18)6-2-14(1)9-15-3-7-17(8-4-15)21-11-19-13-23-19/h1-8,18-19H,9-13H2 |
| SMILES | C(Oc1ccc(Cc2ccc(OCC3CO3)cc2)cc1)C4CO4 |
| IUPAC Name | 2-[[4-[[4-(oxiran-2-ylmethoxy)phenyl]methyl]phenoxy]methyl]oxirane |
| Molecular Weight (g/mol) | 312.36 |
| Show More |