Learn More
Bisphenol F Bis(2-chloro-1-propanol) Ether(Mixture of Diastereomers), TRC
Specifications
Specifications
| Chemical Name or Material | Bisphenol F Bis(2-chloro-1-propanol) Ether |
| CAS | 374772-79-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19H22Cl2O4 |
| Quantity | 50 mg |
| Synonym | BFDGE.2HCL (Badge 7), Bisphenol F Bis(2-chloro-3-hydroxypropyl) Ether, 3,3'-[Methylenebis(4,1-phenyleneoxy)]bis[2-chloro-1-propanol], 1-Propanol, 3,3'-[methylenebis(4,1-phenyleneoxy)]bis[2-chloro- |
| InChI Formula | InChI=1S/C19H22Cl2O4/c20-16(10-22)12-24-18-5-1-14(2-6-18)9-15-3-7-19(8-4-15)25-13-17(21)11-23/h1-8,16-17,22-23H,9-13H2 |
| SMILES | OCC(Cl)COc1ccc(Cc2ccc(OCC(Cl)CO)cc2)cc1 |
| IUPAC Name | 2-chloro-3-[4-[[4-(2-chloro-3-hydroxypropoxy)phenyl]methyl]phenoxy]propan-1-ol |
| Molecular Weight (g/mol) | 385.28 |
| Show More |