Learn More
Bisphenol A-2,2',3,3',5,5',6,6'-d8, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Bisphenol A ring-d8 |
| CAS | 92739-58-7 |
| Molecular Formula | C15 2H8 H8 O2 |
| Quantity | 0.1 g |
| Synonym | Phen-2,3,5,6-d4-ol, 4,4'-(1-methylethylidene)bis-, 4,4'-(1-Methylethylidene)bis[phen-2,3,5,6-d4-ol], Bisphenol A-d8, Bisphenol A (ring-D8), 2,2-Bis(4-hydroxy-2,3,5,6-tetradeuteriophenyl)propane, 4,4'-Isopropylidenediphenol (ring-D8) |
| InChI Formula | InChI=1 S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17 H,1-2H3/i3D,4 D,5 D,6 D,7 D,8 D,9 D,10 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1O)C(C)(C)c2c([2 H])c([2 H])c(O)c([2 H])c2[2 H] |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-[2-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)propan-2-yl]phenol |
| Molecular Weight (g/mol) | 236.34 |
| Formula Weight | 236.1652 g/mol |
| Show More |