Learn More
Bisoctrizole, TRC
Product Code. 30134344
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134344 50 g 50g
30134884 100 g 100g
17937844 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134344 Supplier Toronto Research Chemicals Supplier No. TRCB50887050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 103597-45-1
Molecular Formula C41 H50 N6 O2
Molecular Weight (g/mol) 658.87
Synonym 2,2'-Methylenebis[6-(2H-benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol], 2,2'-Methylenebis(4-tert-octyl-6-benzotriazol-2-ylphenol), 2,2'-Methylenebis[4-(1,1,3,3-tetramethylbutyl)-6-(2H-benzotriazol-2-yl)phenol], 2,2'-Methylenebis[4-(1,1,3,3-tetramethylbutyl)-6-benzotriazol-2-ylphenol], 2,2'-Methylenebis[4-(1,1,3,3-tetramethylbutyl)-6-benzotriazolylphenol], 2,2'-Methylenebis[6-(2-benzotriazolyl)-4-(1,1,3,3-tetramethylbutyl)phenol], 2,2'-Methylenebis[6-(2H-benzotriazol-2-yl)-4-tert-octylphenol], 2,2'-Methylenebis[6-(2H-benzotriazole-2-yl)-4-tert-octylphenol], ADK Stab LA 31, ADK Stab LA 31R, ADK Stab LA 31RG, ADK-ARKLS LA 31, Bis[2-hydroxy-5-tert-octyl-3-(benzotriazol-2-yl)phenyl]methane, Bisoctrizole, Bisoctyltriazole, Chiguard 5431, Eversorb 78, FAT 75'634, JF 832, Kemisorb 279, LA 31, LA 31RG, MBBT, Mark LA 31, Methylenebis(2-hydroxy-3-(benzotriazol-2-yl)-5-tert-octylphenyl), Mixxim ADK LA 31, Mixxim BB 100, Tinosorb M, Tinuvin 360, UV 360, ZINC 11677911
IUPAC Name 2-(benzotriazol-2-yl)-6-[[3-(benzotriazol-2-yl)-2-hydroxy-5-(2,4,4-trimethylpentan-2-yl)phenyl]methyl]-4-(2,4,4-trimethylpentan-2-yl)phenol
SMILES CC(C)(C)CC(C)(C)c1cc(Cc2cc(cc(c2O)n3nc4ccccc4n3)C(C)(C)CC(C)(C)C)c(O)c(c1)n5nc6ccccc6n5
Specifications
Quantity 50 g
InChI Formula InChI=1S/C41H50N6O2/c1-38(2,3)24-40(7,8)28-20-26(36(48)34(22-28)46-42-30-15-11-12-16-31(30)43-46)19-27-21-29(41(9,10)25-39(4,5)6)23-35(37(27)49)47-44-32-17-13-14-18-33(32)45-47/h11-18,20-23,48-49H,19,24-25H2,1-10H3
Formula Weight 658.3995
Chemical Name or Material Bisoctrizole