Learn More
Biscarbofuran Sulfide, TRC
Specifications
Specifications
| Chemical Name or Material | Biscarbofuran Sulfide |
| CAS | 50673-14-8 |
| Molecular Formula | C24 H28 N2 O6 S |
| Quantity | 25 mg |
| Synonym | Carbosulfan Impurity B |
| InChI Formula | InChI=1S/C24H28N2O6S/c1-23(2)13-15-9-7-11-17(19(15)31-23)29-21(27)25(5)33-26(6)22(28)30-18-12-8-10-16-14-24(3,4)32-20(16)18/h7-12H,13-14H2,1-6H3 |
| SMILES | CN(SN(C)C(=O)Oc1cccc2CC(C)(C)Oc12)C(=O)Oc3cccc4CC(C)(C)Oc34 |
| IUPAC Name | (2,2-dimethyl-3H-1-benzofuran-7-yl) N-[(2,2-dimethyl-3H-1-benzofuran-7-yl)oxycarbonyl-methylamino]sulfanyl-N-methylcarbamate |
| Molecular Weight (g/mol) | 472.554 |
| Formula Weight | 472.167 |