missing translation for 'onlineSavingsMsg'
Learn More

Bisbentiamine, TRC

Product Code. 30132810
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132810 500 mg 500mg
30132865 250 mg 250mg
30005315 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132810 Supplier Toronto Research Chemicals Supplier No. TRCB411138500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2667-89-2
Molecular Formula C38H42N8O6S2
Molecular Weight (g/mol) 770.92
Synonym N,N'-[dithiobis[2-(2-hydroxyethyl)-1-methylvinylene]]bis[N-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-Formamide dibenzoate (ester) (8CI), N,N’-[dithiobis[2-[2-(benzoyloxy)ethyl]-1-methyl-2,1-ethenediyl]]bis[N-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-Formamide (9CI), N-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-N-[2-[[2-[[(4-amino-2-methyl-5-pyrimidinyl)methyl]formylamino]-1-[2-(benzoyloxy)ethyl]-1-propen-1-yl]dithio]-4-(benzoyloxy)-1-methyl-1-buten-1-yl]formamide, BTDS, Benzoylthiamine disulfide, Beprocin, Beston, Bis(O-benzoylthiamine) disulfide, O-Benzoylthiamine disulfide
IUPAC Name [(E)-4-[(4-amino-2-methylpyrimidin-5-yl)methyl-formylamino]-3-[[(E)-2-[(4-amino-2-methylpyrimidin-5-yl)methyl-formylamino]-5-benzoyloxypent-2-en-3-yl]disulfanyl]pent-3-enyl] benzoate
SMILES C\C(=C(\CCOC(=O)c1ccccc1)/SS\C(=C(/C)\N(Cc2cnc(C)nc2N)C=O)\CCOC(=O)c3ccccc3)\N(Cc4cnc(C)nc4N)C=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C38H42N8O6S2/c1-25(45(23-47)21-31-19-41-27(3)43-35(31)39)33(15-17-51-37(49)29-11-7-5-8-12-29)53-54-34(16-18-52-38(50)30-13-9-6-10-14-30)26(2)46(24-48)22-32-20-42-28(4)44-36(32)40/h5-14,19-20,23-24H,15-18,21-22H2,1-4H3,(H2,39,41,43)(H2,40,42,44)/b33-25+,34-26+
Formula Weight 770.2669
Percent Purity >95%
Chemical Name or Material Bisbentiamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.