Learn More
Bis(perfluorohexyl)phosphinic Acid Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Bis(perfluorohexyl)phosphinic acid sodium salt |
| CAS | 70609-44-8 |
| Molecular Formula | C12 F26 O2 P . Na |
| Quantity | 50 mg |
| Synonym | 6:6 PFPi, 6:6 PhiA, Phosphinic acid, bis(tridecafluorohexyl)-, sodium salt (9CI) |
| InChI Formula | InChI=1S/C12HF26O2P.Na/c13-1(14,5(21,22)9(29,30)31)3(17,18)7(25,26)11(35,36)41(39,40)12(37,38)8(27,28)4(19,20)2(15,16)6(23,24)10(32,33)34;/h(H,39,40);/q;+1/p-1 |
| SMILES | [Na+].[O-]P(=O)(C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC Name | sodium;bis(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)phosphinate |
| Molecular Weight (g/mol) | 724.05 |
| Formula Weight | 723.9118 |