Learn More
Bis(P-sulfonatophenyl)phenylphosphine Hydrate Dipotassium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Bis(P-sulfonatophenyl)phenylphosphine Dihydrate Dipotassium Salt |
| CAS | 308103-66-4 |
| Molecular Formula | C18 H13 O6 P S2 . 2[H2O] . 2[K] |
| Quantity | 100 mg |
| Synonym | Dipotassium bis(p-sulfonatophenyl)phenylphosphine dihydrate, Bis(p-sulfonatophenyl)phenyl phosphine dipotassium dihydrate |
| InChI Formula | InChI=1S/C18H15O6PS2.2K.2H2O/c19-26(20,21)17-10-6-15(7-11-17)25(14-4-2-1-3-5-14)16-8-12-18(13-9-16)27(22,23)24;;;;/h1-13H,(H,19,20,21)(H,22,23,24);;;2*1H2/q;2*+1;;/p-2 |
| SMILES | O.O.[K+].[K+].[O-]S(=O)(=O)c1ccc(cc1)P(c2ccccc2)c3ccc(cc3)S(=O)(=O)[O-] |
| IUPAC Name | dipotassium;4-[phenyl-(4-sulfonatophenyl)phosphanyl]benzenesulfonate;dihydrate |
| Molecular Weight (g/mol) | 534.62314 |
| Formula Weight | 533.9377 |