Learn More
Bis(p-nitrobenzyl) Phosphate, TRC
Specifications
Specifications
| Chemical Name or Material | Bis(p-nitrobenzyl) Phosphate |
| CAS | 14390-40-0 |
| Molecular Formula | C14H13N2O8P |
| Quantity | 50 mg |
| Synonym | 4-Nitro-benzenemethanol Hydrogen Phosphate, p-Nitro-benzyl Alcohol Hydrogen Phosphate, NSC 401700 |
| InChI Formula | InChI=1S/C14H13N2O8P/c17-15(18)13-5-1-11(2-6-13)9-23-25(21,22)24-10-12-3-7-14(8-4-12)16(19)20/h1-8H,9-10H2,(H,21,22)/p-1 |
| SMILES | [O-][N+](=O)c1ccc(COP(=O)([O-])OCc2ccc(cc2)[N+](=O)[O-])cc1 |
| IUPAC Name | bis[(4-nitrophenyl)methyl] phosphate |
| Molecular Weight (g/mol) | 368.24 |
| Formula Weight | 367.0331 |