Learn More
Bis(p-acetylaminophenyl) Ether, TRC
Product Code. 30138604
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138604 10 g 10g
17963735 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30138604 Supplier Toronto Research Chemicals Supplier No. TRCB40010010G
 Restricted Item
Estimated Shipment: 24-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3070-86-8
Molecular Formula C16 H16 N2 O3
Molecular Weight (g/mol) 284.31
Synonym N,N'-(oxydi-4,1-phenylene)bisacetamide, Acetanilide, 4',4' ''-oxybis- (6CI,7CI,8CI), N,N'-(Oxydi-4,1-phenylene)bis[acetamide], 4,4'-Bis(acetylamino)diphenyl ether, 4,4'-Diacetamidodiphenyl ether, 4,4'-Oxybis[acetanilide], 4',4' ''-Oxybisacetanilide, Bis(p-acetylaminophenyl) ether, NSC 19584, N,N'-(Oxydibenzene-4,1-diyl)diacetamide, Paracetamol Imp. N (Pharmeuropa)
IUPAC Name N-[4-(4-acetamidophenoxy)phenyl]acetamide
SMILES CC(=O)Nc1ccc(Oc2ccc(NC(=O)C)cc2)cc1
Specifications
Quantity 10 g
InChI Formula InChI=1S/C16H16N2O3/c1-11(19)17-13-3-7-15(8-4-13)21-16-9-5-14(6-10-16)18-12(2)20/h3-10H,1-2H3,(H,17,19)(H,18,20)
Formula Weight 284.1161
Percent Purity >95%
Chemical Name or Material N,N'-(Oxydibenzene-4,1-diyl)diacetamide
Grade HPLC