Learn More
Bis(dibenzylideneacetone)palladium (Technical Grade), TRC
Product Code. 30329846
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30329846 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30329846 Supplier Toronto Research Chemicals Supplier No. TRCB4261552.5G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bis(dibenzylideneacetone)palladium
CAS 32005-36-0
Molecular Formula 2 C17 H14 O . Pd
Quantity 2.5 g
Synonym Bis[(1,2,4,5-η)-1,5-diphenyl-1,4-pentadien-3-one]palladium
InChI Formula InChI=1S/2C17H14O.Pd/c2*18-17(13-11-15-7-3-1-4-8-15)14-12-16-9-5-2-6-10-16;/h2*1-14H;/b2*13-11+,14-12+;
SMILES [Pd].O=C(\C=C\c1ccccc1)\C=C\c2ccccc2.O=C(\C=C\c3ccccc3)\C=C\c4ccccc4
IUPAC Name (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
Molecular Weight (g/mol) 575
Formula Weight 574.1124