Learn More
Bis(4-propylbenzylidene) Propyl Sorbitol, TRC
Specifications
Specifications
| Chemical Name or Material | Bis(4-propylbenzylidene) Propyl Sorbitol |
| CAS | 882073-43-0 |
| Molecular Formula | C29 H40 O6 |
| Quantity | 10 mg |
| Synonym | 1,2,3-Trideoxy-4,6:5,7-bis-O-[(4-propylphenyl)methylene]nonitol, 1,2,3-Tridesoxy-4,6:5,7-bis-O-[(4-propylphenyl)methylene]nonitol, 8000K, Millad NX 8000, Millad NX 8000J, Millad NX 8000K, NX 8000, NX 8000K |
| InChI Formula | InChI=1S/C29H40O6/c1-4-7-19-10-14-21(15-11-19)28-32-24(9-6-3)26-27(35-28)25(23(31)18-30)33-29(34-26)22-16-12-20(8-5-2)13-17-22/h10-17,23-31H,4-9,18H2,1-3H3 |
| SMILES | CCCC1OC(OC2C(OC(OC12)c3ccc(CCC)cc3)C(O)CO)c4ccc(CCC)cc4 |
| IUPAC Name | 1-[8-propyl-2,6-bis(4-propylphenyl)-4,4a,8,8a-tetrahydro-[1,3]dioxino[5,4-d][1,3]dioxin-4-yl]ethane-1,2-diol |
| Molecular Weight (g/mol) | 484.624 |
| Formula Weight | 484.282 |