missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-nitrophenyl) Phenyl Phosphate, TRC

Product Code. 30335792
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335792 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335792 Supplier Toronto Research Chemicals Supplier No. TRCB596460100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis(4-nitrophenyl)phenyl Phosphate
CAS 38873-96-0
Molecular Formula C18 H13 N2 O8 P
Quantity 100 mg
Synonym Phosphoric acid, bis(4-nitrophenyl) phenyl ester, Bis(4-nitrophenyl)phenyl Phosphate
InChI Formula InChI=1S/C18H13N2O8P/c21-19(22)14-6-10-17(11-7-14)27-29(25,26-16-4-2-1-3-5-16)28-18-12-8-15(9-13-18)20(23)24/h1-13H
SMILES [O-][N+](=O)c1ccc(OP(=O)(Oc2ccccc2)Oc3ccc(cc3)[N+](=O)[O-])cc1
IUPAC Name bis(4-nitrophenyl) phenyl phosphate
Molecular Weight (g/mol) 416.2782
Formula Weight 416.041
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.