Learn More
Bis(4-methoxybenzyl) Ketone, TRC
Specifications
Specifications
| Chemical Name or Material | Bis(4-methoxybenzyl) Ketone |
| CAS | 29903-09-1 |
| Molecular Formula | C17 H18 O3 |
| Quantity | 500 mg |
| Synonym | Venlafaxine Impurity 4 |
| InChI Formula | InChI=1S/C17H18O3/c1-19-16-7-3-13(4-8-16)11-15(18)12-14-5-9-17(20-2)10-6-14/h3-10H,11-12H2,1-2H3 |
| SMILES | COc1ccc(CC(=O)Cc2ccc(OC)cc2)cc1 |
| IUPAC Name | 1,3-bis(4-methoxyphenyl)propan-2-one |
| Molecular Weight (g/mol) | 270.323 |
| Formula Weight | 270.126 |