Learn More
Bis(3-methylthien-2-yl)methanone, TRC
Product Code. 30342520
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30342520 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30342520 Supplier Toronto Research Chemicals Supplier No. TRCB497230100MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bis(3-methylthien-2-yl)methanone
CAS 30717-55-6
Molecular Formula C11H10OS2
Quantity 100 mg
Synonym Bis(3-methyl-2-thienyl) Ketone, Bis(3-methyl-2-thienyl)methanone
InChI Formula InChI=1S/C11H10OS2/c1-7-3-5-13-10(7)9(12)11-8(2)4-6-14-11/h3-6H,1-2H3
SMILES Cc1ccsc1C(=O)c2sccc2C
IUPAC Name bis(3-methylthiophen-2-yl)methanone
Molecular Weight (g/mol) 222.33
Formula Weight 222.0173