Learn More
Bis(3-methylthien-2-yl)methanone, TRC
Product Code. 30339965
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30339965 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30339965 Supplier Toronto Research Chemicals Supplier No. TRCB4972301G
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bis(3-methylthien-2-yl)methanone
CAS 30717-55-6
Molecular Formula C11H10OS2
Quantity 1 g
Synonym Bis(3-methyl-2-thienyl) Ketone, Bis(3-methyl-2-thienyl)methanone
InChI Formula InChI=1S/C11H10OS2/c1-7-3-5-13-10(7)9(12)11-8(2)4-6-14-11/h3-6H,1-2H3
SMILES Cc1ccsc1C(=O)c2sccc2C
IUPAC Name bis(3-methylthiophen-2-yl)methanone
Molecular Weight (g/mol) 222.33
Formula Weight 222.0173